missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Thiocyanate, Certified, 30.0% (w/v) ±0.5% (w/v), LabChem™
Specifications
Specifications
| Color | Colorless |
| CAS Max % | 70, 30 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Linear Formula | NH4SCN |
| Solubility Information | Soluble in water |
| Formula Weight | 76.12 |
| Percent Purity | 30.0% ±0.5% (w/v) |
| Grade | Certified |
| Identification | Passes Test |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Harmful if inhaled. Harmful to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Avoid breathing mist, vapors, spray. Use only outdoors or in a well-ventilated area. Avoid release to the environment. If inhaled: Remove person to fresh air and keep comfortable for breathing. Call a poison center/doctor if you feel unwell. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |