missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Thiocyanate, Certified, 0.100N ±0.001N (0.1M), LabChem™

Catalog No. LC113702
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC113702 1 L
LC113704 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC113702 Supplier LabChem Supplier No. LC113702

Chemical Identifiers

CAS 1762-95-4 , 7732-18-5
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.117
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
PubChem CID 15666
IUPAC Name azanium;thiocyanate
SMILES C(#N)[S-].[NH4+]

Specifications

Color Colorless
CAS Max % 99.24, 0.76
Physical Form Liquid
Quantity 1 L
Linear Formula NH4SCN
Solubility Information Soluble in water
Formula Weight 76.12
Grade Certified
Concentration or Composition (by Analyte or Components) 0.100N ±0.001N (0.1M)
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Gaseous Ammonia; Sulfur compounds; Nitrogen oxides; Carbon monoxide; Carbon dioxide
Chemical Name or Material Ammonium Thiocyanate
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature