missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Thiocyanate, Certified, 0.0250N ±0.0004N (0.025M), LabChem™

Catalog No. LC113501
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC113501 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC113501 Supplier LabChem Supplier No. LC113501

Chemical Identifiers

CAS 1762-95-4 , 7732-18-5
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.117
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
PubChem CID 15666
IUPAC Name azanium;thiocyanate
SMILES C(#N)[S-].[NH4+]

Specifications

Color Colorless
CAS Max % 99.81, 0.19
Physical Form Liquid
Quantity 500 mL
Linear Formula NH4SCN
Solubility Information Soluble in water
Formula Weight 76.12
Grade Certified
Concentration or Composition (by Analyte or Components) 0.0250N ±0.0004N (0.025M)
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Gaseous Ammonia; Sulfur compounds; Nitrogen oxides; Carbon monoxide; Carbon dioxide
Chemical Name or Material Ammonium Thiocyanate
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature