Learn More
Ammonium thiocyanate, 98+%
CAS: 1762-95-4 | CH4N2S | 76.117 g/mol
Supplier: Thermo Scientific Chemicals A106320I
Description
Ammonium thiocyanate has many uses in manufacturing chemicals such as metal thiocyanates [CuSCN, Ca(SCN)2, NaSCN, KSCN] and in a number of pharmaceuticals. It is used in freezing solutions, pickling, printing, and as a corrosion inhibitor against acid gases. It is used in the photography industry as a stabilizer or an accelerator. It is also used in fertilizers, fabric dyeing, zinc coating, electroplating and as a separator of zirconium and hafnium as well as gold and iron. It is also used in analytical chemistry for titrimetric analysis and to determine the iron content by colorimetry in soft drinks.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAmmonium thiocyanate has many uses in manufacturing chemicals such as metal thiocyanates [CuSCN, Ca(SCN)2, NaSCN, KSCN] and in a number of pharmaceuticals. It is used in freezing solutions, pickling, printing, and as a corrosion inhibitor against acid gases. It is used in the photography industry as a stabilizer or an accelerator. It is also used in fertilizers, fabric dyeing, zinc coating, electroplating and as a separator of zirconium and hafnium as well as gold and iron. It is also used in analytical chemistry for titrimetric analysis and to determine the iron content by colorimetry in soft drinks.
Solubility
Soluble in water, ethanol, methanol, ammonia and acetone. Insoluble in chloroform and ethyl acetate.
Notes
Light sensitive and hygroscopic. Incompatible with strong oxidizing agents, strong acids and lead nitrate.
Specifications
| Ammonium thiocyanate | |
| 147°C to 149°C | |
| Odorless | |
| NH4SCN | |
| 5000 g | |
| Hygroscopic; Light sensitive | |
| ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt | |
| SOIFLUNRINLCBN-UHFFFAOYSA-N | |
| azanium;thiocyanate | |
| 15666 | |
| ≥98% |
| 1762-95-4 | |
| 1.305 | |
| CH4N2S | |
| MFCD00011428 | |
| 3595135 | |
| 14,561 | |
| Soluble in water,ethanol,methanol,ammonia and acetone. Insoluble in chloroform and ethyl acetate. | |
| C(#N)[S-].[NH4+] | |
| 76.117 | |
| 76.12 |
Chemical Identifiers
| 1762-95-4 | |
| 76.117 | |
| SOIFLUNRINLCBN-UHFFFAOYSA-N | |
| 15666 | |
| C(#N)[S-].[NH4+] |
| CH4N2S | |
| MFCD00011428 | |
| ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt | |
| azanium;thiocyanate |
Safety and Handling
GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P363-P501c
H302+H312+H332-H318
EINECSNumber : 217-175-6
RTECSNumber : XK7875000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only