missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Thiocyanate, 0.0250 N (N/40), Ricca Chemical

Supplier:  Ricca Chemical Company R069470020F

Quantity 20 L
Packaging Cubitainer

 View more versions of this product

Catalog No. R069470020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

1762-95-4 , 7732-18-5
76.117
ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
azanium;thiocyanate
CH4N2S
SOIFLUNRINLCBN-UHFFFAOYSA-N
15666
C(#N)[S-].[NH4+]

Specifications

Ammonium Thiocyanate
98.0, 0.19
CH4N2S
SOIFLUNRINLCBN-UHFFFAOYSA-N
azanium;thiocyanate
15666
20 L
1762-95-4 , 7732-18-5
100.0, 0.19
ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
C(#N)[S-].[NH4+]
76.117
Cubitainer
0.025Normal
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents