missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tetrathiotungstate

Catalog No. AA2237304
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2237304 2 g
AA2237309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA2237304 Supplier Thermo Scientific Chemicals Supplier No. 02237304
Only null left

CAS: 13862-78-7 | H8N2S4W-2 | 348.158 g/mol

Ammonium tetrathiotungstate is used to classic building block for transition metal sulfide cluster compounds was recently used in a low-temperature, solid state synthesis of a new series of heterometallic sulfide clusters stabilized with dppm [bis(diphenylphosphino)methane] ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13862-78-7
Molecular Formula H8N2S4W-2
Molecular Weight (g/mol) 348.158
MDL Number MFCD00061410
InChI Key CCFCMIWYHMKPAO-UHFFFAOYSA-L
Synonym ammonium tetrathiotungstate, ammonium tetrathiotungstate vi, h2s4w.2nh3, disulfanylidenetungstendithiol diamine, ammonium tetrathiotungstate trace metals basis
PubChem CID 53393497
IUPAC Name azane;bis(sulfanylidene)tungsten;sulfanide
SMILES N.N.[SH-].[SH-].S=[W]=S

Specifications

Color Yellow
Physical Form Chunks and/or Powder
Quantity 2 g
Solubility Information Insoluble in water.
Formula Weight 348.07
Odor Like rotten eggs
Sensitivity Hygroscopic
Chemical Name or Material Ammonium tetrathiotungstate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only