CAS RN 13862-78-7
IUPAC Name:
disulfanylidenetungsten diamine disulfanide
Synonyms:
tungsten disulfide diamine disulfanide
Molecular Weight (g/mol):
348.16
Molecular Formula:
H8N2S4W
InChi Key:
CCFCMIWYHMKPAO-UHFFFAOYSA-L
SMILES:
N.N.[SH-].[SH-].S=[W]=S
1
–
1
of
1
results
1
Ammonium tetrathiotungstate
CAS: 13862-78-7 Molecular Formula: H8N2S4W-2 Molecular Weight (g/mol): 348.158 MDL Number: MFCD00061410 InChI Key: CCFCMIWYHMKPAO-UHFFFAOYSA-L Synonym: ammonium tetrathiotungstate,ammonium tetrathiotungstate vi,h2s4w.2nh3,disulfanylidenetungstendithiol diamine,ammonium tetrathiotungstate trace metals basis PubChem CID: 53393497 IUPAC Name: azane;bis(sulfanylidene)tungsten;sulfanide SMILES: N.N.[SH-].[SH-].S=[W]=S