missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tetrachloroaurate(III) hydrate, Premion™, 99.99% (metals basis)

Catalog No. AA4043406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4043406 5 g
AA4043403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4043406 Supplier Thermo Scientific Chemicals Supplier No. 04043406
Only null left

CAS: 13874-04-9 | AuCl4H4N | 356.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13874-04-9
Molecular Formula AuCl4H4N
Molecular Weight (g/mol) 356.82
MDL Number MFCD00011503
InChI Key APXMUHKEDBIQKQ-UHFFFAOYSA-J
Synonym ammonium tetrachloroaurate iii hydrate, ammonium tetrachloroaurate iii hydrate, premion, ammonium tetrachloroaurate ii, azanium;tetrachlorogold 1-;hydrate, ammonium tetrachloroaurate iii n-hydrate, ammonium tetrachloroaurate 1--water 1/1/1, ammoniumtetrachloroaurate iii hydrate-au puratrem, ammonium tetrachloroaurate iii hydrate, premion r metals basis
PubChem CID 16211474
SMILES N.Cl[Au](Cl)(Cl)Cl

Specifications

Color Yellow to Orange
Physical Form Crystalline Powder
Quantity 5 g
UN Number UN3260
Solubility Information Soluble in water and alcohol
IUPAC Name tetrachlorogold amine
Formula Weight 356.82 (Anhydrous)
Percent Purity 99.99%
Grade Premion
Odor Odorless
Assay (metals basis)
Chemical Name or Material Ammonium tetrachloroaurate(III) hydrate
Show More Show Less
Hazard Category H290-H301-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318-H290
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
Precautionary Statement P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 250-476-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only