missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfide, 20-24% aq. soln.

CAS: 12135-76-1 | H8N2S | 68.14 g/mol

$150.58 - $248.90

Chemical Identifiers

CAS 12135-76-1
Molecular Formula H8N2S
Molecular Weight (g/mol) 68.14
MDL Number MFCD00010892
InChI Key UXXGWUQNRMPNKH-UHFFFAOYSA-N
IUPAC Name diamine sulfane
SMILES N.N.S
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA33286AE
View Documents
Thermo Scientific Chemicals
033286AE
100 mL
Each for $150.58
Only null left
 
AA33286AP
View Documents
Thermo Scientific Chemicals
033286AP
500 mL
Each for $248.90
Only null left
 
Description

Description

Ammonium sulfide solution is used occasionally in photographic developing, to apply patina to bronze, and in textile manufacturing. Ammonium sulfide is a very useful reagent in mineral analysis. Used in continuous culture of saccharomyces cerevisiae, useful in a double staining technique for visualization of astrocytes and microglia from brain sections and astroglial cell cultures. It plays an important role to modify the surface of low κ dielectric thin films. It is essential for molecular beam epitaxy regrowth. It is an unstable salt and acts as a selective reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12135-76-1
68.14
UXXGWUQNRMPNKH-UHFFFAOYSA-N
N.N.S
H8N2S
MFCD00010892
diamine sulfane

Specifications

12135-76-1
Liquid
20 to 24% (aqueous)
MFCD00010892
Miscible with water,alcohol and liquid ammonia.
N.N.S
68.14
Like rotten eggs
546 hPa (410mm Hg) at 20°C
Ammonium sulfide
Colorless to Orange
100 mL
H8N2S
32°C (90°F)
UXXGWUQNRMPNKH-UHFFFAOYSA-N
diamine sulfane
68.14
Colorless liquid
0.997 g/cm3 at 20°C
Videos
Safety and Handling

Safety and Handling

When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Use appropriate containment to avoid environmental contamination.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : DOT Class: 8; Packing Group: II

EINECSNumber : 235-223-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only