missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium sulfate, Enzyme grade, ≥99%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0215037391
Specifications
| Ammonium Sulfate | |
| 7783-20-2 | |
| 1 kg | |
| H8N2O4S | |
| ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
| N.N.OS(O)(=O)=O | |
| 132.13 | |
| CHEBI:62946 | |
| 1.77g/cm3 at 25°C |
| White | |
| Crystalline Powder | |
| ≥99% | |
| MFCD00003391 | |
| BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
| sulfuric acid diamine | |
| 6097028 | |
| 132.14 |
Chemical Identifiers
| 7783-20-2 | |
| 132.13 | |
| BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
| 6097028 | |
| sulfuric acid diamine |
| H8N2O4S | |
| MFCD00003391 | |
| ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
| CHEBI:62946 | |
| N.N.OS(O)(=O)=O |
Safety and Handling
Recommended Storage : Store at room temperature.