missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfate, Enzyme grade, ≥99%, MP Biomedicals™

Catalog No. p-4404655
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15037380 100 g
ICN15037391 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15037380 Supplier MP Biomedicals Inc Supplier No. 0215037380

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7783-20-2 | Formula: H8N2O4S | Weight: 132.13 g/mol

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
IUPAC Name sulfuric acid diamine
SMILES N.N.OS(O)(=O)=O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range ≥99%
Density 1.77g/cm3 at 25°C
Formula Weight 132.14
Percent Purity 100.3%
Grade Not Available
Chemical Name or Material Ammonium Sulfate
Recommended Storage Store at room temperature.