missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfate, 0.5M aq. soln.

CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol

Supplier:  Thermo Scientific Chemicals J60368AE

 View more versions of this product

Catalog No. AAJ60368AE


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ammonium sulfate
Liquid
H8N2O4S
MFCD00003391
BFNBIHQBYMNNAN-UHFFFAOYSA-N
diazanium;sulfate
6097028
132.14
7783-20-2
100 mL
(NH4)2SO4
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
N.N.OS(O)(=O)=O
132.13
CHEBI:62946
0.5M aq. soln.

Chemical Identifiers

7783-20-2
132.13
BFNBIHQBYMNNAN-UHFFFAOYSA-N
6097028
diazanium;sulfate
H8N2O4S
MFCD00003391
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
CHEBI:62946
N.N.OS(O)(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only