missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfate, 0.5M aq. soln.

Catalog No. p-7041817
Change view
Click to view available options
Quantity:
50 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60368AD 50 mL
AAJ60368AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60368AD Supplier Thermo Scientific Chemicals Supplier No. J60368AD
Only null left

CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate
PubChem CID 6097028
ChEBI CHEBI:62946
IUPAC Name diazanium;sulfate
SMILES N.N.OS(O)(=O)=O

Specifications

Physical Form Liquid
Quantity 50 mL
Linear Formula (NH4)2SO4
Formula Weight 132.14
Concentration or Composition (by Analyte or Components) 0.5M aq. soln.
Chemical Name or Material Ammonium sulfate
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only