missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium sulfamate, ACS reagent
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 132.0°C to 134.0°C |
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 100 g |
| Linear Formula | H2NSO3NH4 |
| Merck Index | 15, 551 |
| Formula Weight | 114.12 |
| Percent Purity | ≥98.0% |
| Grade | ACS Reagent |
| Packaging | Plastic Bottle |
| Show More |
Safety and Handling
| Hazard Category | HYGROSCOPIC |
| Hazard Statement | GHS H Statement: Harmful if swallowed. |
| Precautionary Statement | GHS P Statement: Wash face, hands and any exposed skin thoroughly after handling. Do not eat, drink or smoke when using this product. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 231-871-7 |
| RTECSNumber | WO6125000 |
| TSCA | TSCA |
RUO – Research Use Only