missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfamate, ACS reagent

Catalog No. AC423390050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423390050 5 g Glass Bottle
AC423391000 100 g Plastic Bottle
AC423395000 500 g Plastic Bottle
AC423390025 2.5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC423390050 Supplier Thermo Scientific Chemicals Supplier No. 423390050

CAS: 7773-06-0 | H6N2O3S | 114.119 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7773-06-0
Molecular Formula H6N2O3S
Molecular Weight (g/mol) 114.119
MDL Number MFCD00011429
InChI Key GEHMBYLTCISYNY-UHFFFAOYSA-N
Synonym ammonium sulfamate, ammonium sulphamate, ammate, ikurin, ammonium amidosulfonate, feliderm k, ammate x, sulfamic acid, monoammonium salt, ammonium sulfamidate, ammonium sulphamidate
PubChem CID 24482
ChEBI CHEBI:81950
IUPAC Name azanium;sulfamate
SMILES [NH4+].NS(=O)(=O)[O-]

Specifications

Melting Point 132.0°C to 134.0°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
Linear Formula H2NSO3NH4
Merck Index 15, 551
Formula Weight 114.12
Percent Purity ≥98.0%
Grade ACS Reagent
Packaging Glass Bottle
Residue after Ignition 0.10% max.
Chemical Name or Material Ammonium sulfamate, 98+%
Show More Show Less
Hazard Category HYGROSCOPIC
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 231-871-7
RTECSNumber WO6125000
TSCA TSCA

RUO – Research Use Only