missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Sulfamate, ACS, 98%, Spectrum™ Chemical
Specifications
Specifications
| Melting Point | Within 2.0°C |
| CAS Max % | 1 |
| Quantity | 100 g |
| Percent Purity | 98% |
| Grade | ACS |
| Packaging | Amber Glass Bottle |
| Residue after Ignition | 0.001 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.