missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfamate, ACS, 98%, Spectrum™ Chemical

Catalog No. 18608779
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
100 lb.
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608779 100 lb. Amber Glass Bottle
18608775 100 g Amber Glass Bottle
18608777 500 g Amber Glass Bottle
18608778 2.5 kg Amber Glass Bottle
18608776 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608779 Supplier Spectrum Chemical Mfg Cor Supplier No. A1243100LBBL
Get Up to $500 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7773-06-0

Specifications

Melting Point Within 2.0°C
CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 100 lb.
Molecular Weight (g/mol) 114.13
Percent Purity 98%
Grade Reagent
Packaging Amber Glass Bottle
Residue after Ignition 0.001
Chemical Name or Material Ammonium Sulfamate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.