missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfamate, ACS, 98%, Spectrum™ Chemical

Catalog No. 18608775
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
100 lb.
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608775 100 g Amber Glass Bottle
18608777 500 g Amber Glass Bottle
18608779 100 lb. Amber Glass Bottle
18608778 2.5 kg Amber Glass Bottle
18608776 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608775 Supplier Spectrum Chemical Mfg Cor Supplier No. A1243100GM

Chemical Identifiers

CAS 7773-06-0

Specifications

Melting Point Within 2.0°C
CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 100 g
Molecular Weight (g/mol) 114.13
Percent Purity 98%
Grade Reagent
Packaging Amber Glass Bottle
Residue after Ignition 0.001
Chemical Name or Material Ammonium Sulfamate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.