missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfamate, ACS, 98% min

Catalog No. p-4407232
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1453436 500 g
AA14534A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1453436 Supplier Thermo Scientific Chemicals Supplier No. 01453436
Only null left

CAS: 7773-06-0 | H6N2O3S | 114.119 g/mol

Ammonium sulfamate is applied in water solution or oil-water emulsion as a leaf, or foliar spray for control of woody plants, or it is applied as crystals or concentrated solution to cuts in the bark or on freshly-cut stumps of undesirable trees to prevent resprouting. It is used as a compost accelerator, flame retardant and in industrial processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7773-06-0
Molecular Formula H6N2O3S
Molecular Weight (g/mol) 114.119
MDL Number MFCD00011429
InChI Key GEHMBYLTCISYNY-UHFFFAOYSA-N
Synonym ammonium sulfamate, ammonium sulphamate, ammate, ikurin, ammonium amidosulfonate, feliderm k, ammate x, sulfamic acid, monoammonium salt, ammonium sulfamidate, ammonium sulphamidate
PubChem CID 24482
ChEBI CHEBI:81950
IUPAC Name azanium;sulfamate
SMILES [NH4+].NS(=O)(=O)[O-]

Specifications

Boiling Point 160°C (decomposition)
Melting Point 131°C to 135°C
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 98% min.
Solubility Information Very soluble in water,liquid ammonia. Slightly soluble in ethanol,glycerol,glycol
Formula Weight 114.12
Grade ACS Reagent
Sensitivity Hygroscopic
Chemical Name or Material Ammonium sulfamate, For ACS analysis
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 231-871-7
RTECSNumber WO6125000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only