missing translation for 'onlineSavingsMsg'
Learn More

Ammonium sulfamate, 98+%

Catalog No. p-7023794
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1769630 250 g
AAA176960B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1769630 Supplier Thermo Scientific Chemicals Supplier No. A1769630

CAS: 7773-06-0 | H6N2O3S | 114.119 g/mol

Ammonium sulfamate is applied in water solution or oil-water emulsion as a leaf, or foliar spray for control of woody plants, or it is applied as crystals or concentrated solution to cuts in the bark or on freshly-cut stumps of undesirable trees to prevent resprouting. Used as a compost accelerator, flame retardant and in industrial processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium sulfamate is applied in water solution or oil-water emulsion as a leaf, or foliar spray for control of woody plants, or it is applied as crystals or concentrated solution to cuts in the bark or on freshly-cut stumps of undesirable trees to prevent resprouting. Used as a compost accelerator, flame retardant and in industrial processes.

Solubility
Soluble in water(2800g/L ). Insoluble in methanol, ether, n-octanol.

Notes
Hygroscopic. Store at room temperature. Stored in airtight containers which are kept closed and away from air and moisture.

Chemical Identifiers

CAS 7773-06-0
Molecular Formula H6N2O3S
Molecular Weight (g/mol) 114.119
MDL Number MFCD00011429
InChI Key GEHMBYLTCISYNY-UHFFFAOYSA-N
Synonym ammonium sulfamate, ammonium sulphamate, ammate, ikurin, ammonium amidosulfonate, feliderm k, ammate x, sulfamic acid, monoammonium salt, ammonium sulfamidate, ammonium sulphamidate
PubChem CID 24482
ChEBI CHEBI:81950
IUPAC Name azanium;sulfamate
SMILES [NH4+].NS(=O)(=O)[O-]

Specifications

Boiling Point 160°C (decomposition)
Melting Point 131°C to 135°C
pH 5.2
Quantity 250 g
Linear Formula NH4SO3NH2
Merck Index 14,554
Solubility Information Soluble in water(2800g/L ). Insoluble in methanol,ether,n-octanol.
Formula Weight 114.12
Percent Purity ≥98%
Odor Weak
Sensitivity Hygroscopic
Chemical Name or Material Ammonium sulfamate
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 231-871-7
RTECSNumber WO6125000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only