missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfamate, ≥98.0%, ACS Reagent, Honeywell Fluka™

ACS Reagent,≥98.0%

$500.58 - $500.58

Chemical Identifiers

CAS 7773-06-0
Molecular Formula H6N2O3S
Molecular Weight (g/mol) 114.119
MDL Number MFCD00011429
InChI Key GEHMBYLTCISYNY-UHFFFAOYSA-N
Synonym ammonium sulfamate, ammonium sulphamate, ammate, ikurin, ammonium amidosulfonate, feliderm k, ammate x, sulfamic acid, monoammonium salt, ammonium sulfamidate, ammonium sulphamidate
PubChem CID 24482
ChEBI CHEBI:81950
IUPAC Name azanium;sulfamate
SMILES [NH4+].NS(=O)(=O)[O-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60045402
View Documents
Honeywell Chemicals
228745500G
500 g Poly Bottle N/A
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7773-06-0
114.119
GEHMBYLTCISYNY-UHFFFAOYSA-N
24482
azanium;sulfamate
H6N2O3S
MFCD00011429
ammonium sulfamate, ammonium sulphamate, ammate, ikurin, ammonium amidosulfonate, feliderm k, ammate x, sulfamic acid, monoammonium salt, ammonium sulfamidate, ammonium sulphamidate
CHEBI:81950
[NH4+].NS(=O)(=O)[O-]

Specifications

131°C to 135°C
White
H6N2O3S
MFCD00011429
ammonium sulfamate, ammonium sulphamate, ammate, ikurin, ammonium amidosulfonate, feliderm k, ammate x, sulfamic acid, monoammonium salt, ammonium sulfamidate, ammonium sulphamidate
[NH4+].NS(=O)(=O)[O-]
114.119
CHEBI:81950
≥98.0%
Poly Bottle
Crystalline
Ammonium Sulfamate
7773-06-0
500 g
NH4SO3NH2
UN3077
GEHMBYLTCISYNY-UHFFFAOYSA-N
azanium;sulfamate
24482
114.12g/mol
ACS Reagent
≤0.10%
1g/cm3
Videos
Safety and Handling

Safety and Handling

P273

H302-H400

DOTInformation : 9/PGIII

EINECSNumber : 231-871-7

RTECSNumber : WO6125000

SDS
missing translation for 'documents'

missing translation for 'documents'