missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Phosphate, For Sulfide, 40%, LabChem™
Supplier: LabChem LC112951
Specifications
| Ammonium Phosphate | |
| For Sulfide | |
| 60,40 | |
| (NH4)2HPO4 | |
| diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate | |
| MNNHAPBLZZVQHP-UHFFFAOYSA-N | |
| phosphoric acid diamine | |
| 24540 | |
| 132.06 | |
| Passes Test | |
| Gaseous Ammonia; Phosphorus oxides | |
| Liquid |
| Colorless | |
| 7783-28-0,7732-18-5 | |
| H9N2O4P | |
| MFCD00010891 | |
| Soluble in water | |
| N.N.OP(O)(O)=O | |
| 132.06 | |
| CHEBI:63051 | |
| 40% | |
| Poly Bottle | |
| 500 mL |
Chemical Identifiers
| 7783-28-0 | |
| 132.06 | |
| MNNHAPBLZZVQHP-UHFFFAOYSA-N | |
| 24540 | |
| phosphoric acid diamine |
| H9N2O4P | |
| MFCD00010891 | |
| diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate | |
| CHEBI:63051 | |
| N.N.OP(O)(O)=O |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature