missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Phosphate Dibasic, puriss., Honeywell Fluka™
Puriss., meets analytical specification of NF, 99-102%
Supplier: Honeywell-Fluka 0421125KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Ammonium Phosphate Dibasic | |
| 7783-28-0 | |
| H9N2O4P | |
| MFCD00010891 | |
| diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate | |
| N.N.OP(O)(O)=O | |
| 132.06 | |
| CHEBI:63051 | |
| 99 to 102% |
| Meets analytical specification of NF | |
| 25 kg | |
| (NH4)2HPO4 | |
| NONH for all modes of transport | |
| MNNHAPBLZZVQHP-UHFFFAOYSA-N | |
| diazanium;hydrogen phosphate | |
| 24540 | |
| 132.06g/mol | |
| puriss. |
Chemical Identifiers
| 7783-28-0 | |
| 132.06 | |
| MNNHAPBLZZVQHP-UHFFFAOYSA-N | |
| 24540 | |
| diazanium;hydrogen phosphate |
| H9N2O4P | |
| MFCD00010891 | |
| diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate | |
| CHEBI:63051 | |
| N.N.OP(O)(O)=O |
Safety and Handling
EINECSNumber : 231-987-8