missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate Dibasic, puriss., Honeywell Fluka™

Catalog No. p-7071714
Change view
Click to view available options
Quantity:
1 kg
25 kg
6 x 1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001824 1 kg
60044538 6 x 1 kg
6001825 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6001824 Supplier Honeywell Chemicals Supplier No. 042111KG
Only null left

Puriss., meets analytical specification of NF, 99-102%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7783-28-0
Molecular Formula H9N2O4P
Molecular Weight (g/mol) 132.06
MDL Number MFCD00010891
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
Synonym diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
PubChem CID 24540
ChEBI CHEBI:63051
IUPAC Name diazanium;hydrogen phosphate
SMILES N.N.OP(O)(O)=O

Specifications

Quantity 1 kg
Linear Formula (NH4)2HPO4
UN Number NONH for all modes of transport
Formula Weight 132.06g/mol
Percent Purity 99 to 102%
Grade puriss.
Chemical Name or Material Ammonium Phosphate Dibasic, Meets analytical specification of NF
EINECSNumber 231-987-8