Learn More
Ammonium phosphate, dibasic, 98+%, ACS reagent
CAS: 7783-28-0 | H9N2O4P | 132.06 g/mol
Supplier: Thermo Scientific Chemicals 423370025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Ammonium phosphate, dibasic | |
| White | |
| Crystalline Powder, Crystals and/or Chunks | |
| 98+% | |
| (NH4)2HPO4 | |
| 15, 540 | |
| Solubility in water: 58g/100mL (20°C). Other solubilities: practically insoluble in alcohol and acetone | |
| N.N.OP(O)(O)=O | |
| 132.06 | |
| CHEBI:63051 | |
| ≥98% | |
| Plastic bottle |
| 7783-28-0 | |
| 7.7 to 8.1 (5% soln. at 25°C) | |
| 2.5 kg | |
| H9N2O4P | |
| MFCD00010891 | |
| diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate | |
| MNNHAPBLZZVQHP-UHFFFAOYSA-N | |
| phosphoric acid diamine | |
| 24540 | |
| 132.06 | |
| ACS Reagent | |
| 1.61 |
Chemical Identifiers
| 7783-28-0 | |
| 132.06 | |
| MNNHAPBLZZVQHP-UHFFFAOYSA-N | |
| 24540 | |
| N.N.OP(O)(O)=O |
| H9N2O4P | |
| MFCD00010891 | |
| diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate | |
| CHEBI:63051 |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
EINECSNumber : 231-987-8
TSCA : TSCA
RUO – Research Use Only