missing translation for 'onlineSavingsMsg'
Learn More

Ammonium phosphate, dibasic, 98+%, ACS reagent

Catalog No. p-45630
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423375000 500 g Plastic bottle
AC423370025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC423375000 Supplier Thermo Scientific Chemicals Supplier No. 423375000
Only null left

CAS: 7783-28-0 | H9N2O4P | 132.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7783-28-0
Molecular Formula H9N2O4P
Molecular Weight (g/mol) 132.06
MDL Number MFCD00010891
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
Synonym diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
PubChem CID 24540
ChEBI CHEBI:63051
SMILES N.N.OP(O)(O)=O

Specifications

Color White
pH 7.7 to 8.1 (5% soln. at 25°C)
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 500 g
Assay Percent Range 98+%
Linear Formula (NH4)2HPO4
Merck Index 15, 540
IUPAC Name diazanium;hydrogen phosphate
Formula Weight 132.06
Percent Purity ≥98.0%
Grade ACS Reagent
Packaging Plastic bottle
Density 1.61
Chemical Name or Material Ammonium phosphate, dibasic
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 231-987-8
TSCA TSCA

RUO – Research Use Only