missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Persulfate, ACS Reagent (Ammonium Peroxydisulfate), Reagents
Ammonium Persulfate, ACS Reagent (Ammonium Peroxydisulfate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.
$318.57 - $5106.00
Chemical Identifiers
| CAS | 7727-54-0 |
|---|---|
| Molecular Formula | H8N2O8S2 |
| Molecular Weight (g/mol) | 228.19 |
| InChI Key | ROOXNKNUYICQNP-UHFFFAOYSA-N |
| Synonym | Ammonium Peroxydisulfate |
| IUPAC Name | diammonium [(sulfonatoperoxy)sulfonyl]oxidanide |
| SMILES | [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-338-088
|
Labchem Inc
C1136500100A6 |
100 mL |
Each for $318.57
|
|
|||||
|
01-338-087
|
Labchem Inc
C113650012KG |
12 kg |
Each for $5,106.00
|
|
|||||
|
01-338-086
|
Labchem Inc
C113650025A5 |
2.5 kg |
Each for $1,663.29
|
|
|||||
|
01-338-085
|
Labchem Inc
C1136500500A5 |
500 g |
Each for $508.43
|
|
|||||
Chemical Identifiers
| 7727-54-0 | |
| 228.19 | |
| Ammonium Peroxydisulfate | |
| [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O |
| H8N2O8S2 | |
| ROOXNKNUYICQNP-UHFFFAOYSA-N | |
| diammonium [(sulfonatoperoxy)sulfonyl]oxidanide |
Specifications
| 120°C to 160°C | |
| 7727-54-0 | |
| 100 mL | |
| Ammonium Peroxydisulfate | |
| [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O | |
| 228.19 | |
| Poly Bottle | |
| Ammonium Persulfate, ACS Reagent (Ammonium Peroxydisulfate) |
| Colorless/White | |
| Solid | |
| H8N2O8S2 | |
| ROOXNKNUYICQNP-UHFFFAOYSA-N | |
| diammonium [(sulfonatoperoxy)sulfonyl]oxidanide | |
| ACS | |
| 1.98 |