missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Persulfate, ACS Reagent (Ammonium Peroxydisulfate), Reagents
Ammonium Persulfate, ACS Reagent (Ammonium Peroxydisulfate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C113650012KG
Specifications
| Ammonium Persulfate, ACS Reagent (Ammonium Peroxydisulfate) | |
| Colorless/White | |
| Solid | |
| H8N2O8S2 | |
| ROOXNKNUYICQNP-UHFFFAOYSA-N | |
| diammonium [(sulfonatoperoxy)sulfonyl]oxidanide | |
| ACS | |
| 1.98 |
| 120°C to 160°C | |
| 7727-54-0 | |
| 12 kg | |
| Ammonium Peroxydisulfate | |
| [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O | |
| 228.19 | |
| Poly Pail |
Chemical Identifiers
| 7727-54-0 | |
| 228.19 | |
| Ammonium Peroxydisulfate | |
| [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O |
| H8N2O8S2 | |
| ROOXNKNUYICQNP-UHFFFAOYSA-N | |
| diammonium [(sulfonatoperoxy)sulfonyl]oxidanide |