missing translation for 'onlineSavingsMsg'
Learn More

Ammonium persulfate, 98+%, ACS reagent

Oxidation reagent | CAS: 7727-54-0 | H8N2O8S2 | 228.19 g/mol

Supplier:  Thermo Scientific Chemicals 401165000

 View more versions of this product

Catalog No. AC401165000

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Oxidation reagent
Specifications

Specifications

Ammonium persulfate
7727-54-0
Crystalline Powder and/or Crystals
98+%
(NH4)2S2O8
01,952; 02,348; 03,238; 05,15; 06,20; 12,33
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
228.19
228.19
ACS Reagent
0.05% max.
Negligible
1.9800g/mL
120.0°C
White
500 g
H8N2O8S2
MFCD00003390
15, 539
ROOXNKNUYICQNP-UHFFFAOYSA-N
diazanium;sulfonatooxy sulfate
62648
≥98%
Plastic bottle
1.98
>120°C

Chemical Identifiers

7727-54-0
228.19
ROOXNKNUYICQNP-UHFFFAOYSA-N
62648
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
H8N2O8S2
MFCD00003390
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
diazanium;sulfonatooxy sulfate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful if swallowed.
May intensify fire; oxidizer.

GHS P Statement:
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only