missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Peroxodisulfate, MilliporeSigma™
For analysis EMSURE™ ACS, Reag. Ph Eur
Supplier: Technidata 1012015000
Specifications
| Ammonium peroxodisulfate | |
| 5 kg | |
| MFCD00003390 | |
| ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french | |
| ROOXNKNUYICQNP-UHFFFAOYSA-N | |
| diammonium [(sulfonatoperoxy)sulfonyl]oxidanide | |
| 62648 | |
| Odorless | |
| 1.98g/cm3 at 20°C |
| 7727-54-0 | |
| H8N2O8S2 | |
| UN1444 | |
| Soluble in water: 620g/L at 20°C | |
| [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O | |
| 228.19 | |
| 228.19 | |
| Plastic Bottle |
Chemical Identifiers
| 7727-54-0 | |
| 228.19 | |
| ROOXNKNUYICQNP-UHFFFAOYSA-N | |
| 62648 | |
| [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O |
| H8N2O8S2 | |
| MFCD00003390 | |
| ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french | |
| diammonium [(sulfonatoperoxy)sulfonyl]oxidanide |
Safety and Handling
H302: Harmful if swallowed.
H315: Causes skin irritation.
H272: May intensify fire; oxidiser.
DOTInformation : DOT Class: 5.1, Packaging Group: III
EINECSNumber : 231-786-5
RTECSNumber : SE0350000
Recommended Storage : Store at 15° to 25°C (59° to 77°F), Tightly closed. Dry. Away from combustible substances