missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Peroxodisulfate, MilliporeSigma™

Catalog No. p-5547612
Change view
Click to view available options
Quantity:
500 g
5 kg
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1012010500 500 g Plastic Bottle
M1012015000 5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1012010500 Supplier Technidata Supplier No. 1.01201.0500
Only null left

For analysis EMSURE™ ACS, Reag. Ph Eur

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
PubChem CID 62648
IUPAC Name diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O

Specifications

Quantity 500 g
UN Number UN1444
Solubility Information Soluble in water: 620g/L at 20°C
Formula Weight 228.19
Odor Odorless
Packaging Plastic Bottle
Density 1.98g/cm3 at 20°C
Chemical Name or Material Ammonium peroxodisulfate
Hazard Category H272: May intensify fire; oxidiser.
Hazard Statement H302: Harmful if swallowed.
Precautionary Statement H315: Causes skin irritation.
DOTInformation DOT Class: 5.1, Packaging Group: III
EINECSNumber 231-786-5
RTECSNumber SE0350000
Recommended Storage Store at 15° to 25°C (59° to 77°F), Tightly closed. Dry. Away from combustible substances