missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Oxalate, TS, 0.5 Normal (N/2), Reagents

Catalog No. p-200021857
Change view
Click to view available options
Quantity:
500 mL
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335936 10 L
01335935 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01335936 Supplier Labchem Inc Supplier No. CA10320010F
Only null left

Ammonium Oxalate, TS, 0.5 Normal (N/2), Meets USP specifications, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 6009-70-7
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Diammonium ethanedioic Acid monohydrate
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
Physical Form Liquid
Quantity 10 L
Grade USP
Packaging Cubitainer
Specific Gravity 1.03
Chemical Name or Material Ammonium Oxalate, TS, 0.5 Normal (N/2)