missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Oxalate, TS, 0.5 Normal (N/2), Reagents
Specifications
Specifications
| Boiling Point | 100°C |
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 10 L |
| Grade | USP |
| Packaging | Cubitainer |
| Specific Gravity | 1.03 |
| Chemical Name or Material | Ammonium Oxalate, TS, 0.5 Normal (N/2) |