Learn More
Ammonium oxalate monohydrate, ACS, 99.0-101.0%
In detinning of iron, metal polishing | CAS: 6009-70-7 | C2H10N2O5 | 142.111 g/mol
Supplier: Thermo Scientific Chemicals 03622836
Description
In detinning of iron, metal polishing, determination of lead, calcium, and rare earth metalsAmmonium oxalate monohydrate is widely utilized as a buffering reagent and a dispersant to determine the surface interaction of platelets. It finds an application to study
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- In detinning of iron, metal polishing, determination of lead, calcium, and rare earth metals
Specifications
| Ammonium oxalate | |
| Monohydrate | |
| Granules | |
| 99.0-101.0% | |
| MFCD00149694 | |
| 14,537 | |
| Soluble in water; Slightly soluble in alcohol. | |
| C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O | |
| 142.111 | |
| 142.11 (124.10anhydrous) | |
| ACS |
| 70°C (decomposition) | |
| 6009-70-7 | |
| 500 g | |
| C2H10N2O5 | |
| 3759830 | |
| ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate | |
| MSMNVXKYCPHLLN-UHFFFAOYSA-N | |
| diazanium;oxalate;hydrate | |
| 516808 | |
| 99.0 to 101.0% | |
| 1.5 |
Chemical Identifiers
| 6009-70-7 | |
| 142.111 | |
| MSMNVXKYCPHLLN-UHFFFAOYSA-N | |
| 516808 | |
| C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O |
| C2H10N2O5 | |
| MFCD00149694 | |
| ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate | |
| diazanium;oxalate;hydrate |
Safety and Handling
GHS H Statement
H302-H312
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312-H320-H335-H372
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber : 214-202-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only