missing translation for 'onlineSavingsMsg'
Learn More

Ammonium oxalate monohydrate, ACS, 99.0-101.0%

Catalog No. p-7050226
Change view
Click to view available options
Quantity:
100 g
500 g
4 x 500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3622822 100 g
AA3622836 500 g
AA3622857 4 x 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3622822 Supplier Thermo Scientific Chemicals Supplier No. 03622822
Only null left

In detinning of iron, metal polishing | CAS: 6009-70-7 | C2H10N2O5 | 142.111 g/mol

In detinning of iron, metal polishing, determination of lead, calcium, and rare earth metalsAmmonium oxalate monohydrate is widely utilized as a buffering reagent and a dispersant to determine the surface interaction of platelets. It finds an application to study

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In detinning of iron, metal polishing, determination of lead, calcium, and rare earth metals

Chemical Identifiers

CAS 6009-70-7
Molecular Formula C2H10N2O5
Molecular Weight (g/mol) 142.111
MDL Number MFCD00149694
InChI Key MSMNVXKYCPHLLN-UHFFFAOYSA-N
Synonym ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
PubChem CID 516808
IUPAC Name diazanium;oxalate;hydrate
SMILES C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O

Specifications

Melting Point 70°C (decomposition)
Physical Form Granules
Quantity 100 g
Assay Percent Range 99.0-101.0%
Beilstein 3759830
Merck Index 14,537
Solubility Information Soluble in water; Slightly soluble in alcohol.
Formula Weight 142.11 (124.10 Anhydrous)
Percent Purity 99.0 to 101.0%
Grade ACS
Density 1.5
Chemical Name or Material Ammonium Oxalate Monohydrate, Monohydrate
Show More Show Less
Hazard Category H302+H312-H320-H335-H372
Hazard Statement GHS H Statement
H302-H312
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 214-202-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only