Learn More
Ammonium Molybdate Reagent II, For Phosphorus, LabChem™
$135.30 - $195.87
Chemical Identifiers
| CAS | 12054-85-2 |
|---|---|
| Molecular Formula | H6MoNO2 |
| Molecular Weight (g/mol) | 148.00 |
| InChI Key | XCOMAVCHADVANQ-UHFFFAOYSA-O |
| Synonym | Molybdic acid ammonium salt tetrahydrate, Hexammonium heptamolybdate 4-hydrate |
| PubChem CID | 3084102 |
| IUPAC Name | ammonium hydrate molybdenum oxidandiide |
| SMILES | [NH4+].O.[O--].[Mo] |
Chemical Identifiers
| 12054-85-2 | |
| 148.00 | |
| Molybdic acid ammonium salt tetrahydrate, Hexammonium heptamolybdate 4-hydrate | |
| ammonium hydrate molybdenum oxidandiide |
| H6MoNO2 | |
| XCOMAVCHADVANQ-UHFFFAOYSA-O | |
| 3084102 | |
| [NH4+].O.[O--].[Mo] |
Specifications
| Colorless | |
| 5.08,77.86,17.06 | |
| (NH4)6Mo7O24·4H2O | |
| 1.1g/mL | |
| Soluble in water | |
| [NH4+].O.[O--].[Mo] | |
| 148.00 | |
| 1235.86 | |
| Poly Bottle | |
| 1.3cSt | |
| 500 mL | |
| Ammonium Molybdate Reagent II, For Phosphorus |
| 12054-85-2,7732-18-5,7664-93-9 | |
| H6MoNO2 | |
| UN2796 | |
| Molybdic acid ammonium salt tetrahydrate, Hexammonium heptamolybdate 4-hydrate | |
| XCOMAVCHADVANQ-UHFFFAOYSA-O | |
| ammonium hydrate molybdenum oxidandiide | |
| 3084102 | |
| Passes Test | |
| 1.1g/mL | |
| Sulfur compounds; Fume; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors | |
| Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature