Learn More
Ammonium molybdate (para) tetrahydrate, ACS, 81-83% as MoO{3}
Ammonium molybdate (para) tetrahydrate, H24Mo7N6O24.4H2O, CAS Number-12054-85-2, Ammonium paramolybdate, 2kg, Powder, 81-83%, 2.498, 234-722-4, 1235.86 (1163.79 Anhydrous), ACS, H302-H315-H319-H335, P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c | CAS: 12054-85-2 | H24Mo7N6O24·4H2O
Supplier: Thermo Scientific Chemicals 01183122
Description
Ammonium molybdate (para) tetrahydrate, is used in photography,analytical lab reagent for phosphate,lead,arsenate determination.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Ammonium molybdate (para) tetrahydrate | |
| (para) tetrahydrate | |
| ∽190°C (decomposition) | |
| H24Mo7N6O24·4H2O | |
| 2.498 g/cm3 | |
| Soluble in water (1:2 ratio). Insoluble in alcohol | |
| ACS | |
| 100 g |
| Powder | |
| 12054-85-2 | |
| 81-83% | |
| MFCD00167059 | |
| Ammonium paramolybdate | |
| 1235.86 (1163.79 Anhydrous) | |
| Odorless | |
| Powder |
Chemical Identifiers
| 12054-85-2 | |
| H24Mo7N6O24·4H2O | |
| MFCD00167059 | |
| Ammonium paramolybdate |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 234-722-4
RTECSNumber : QA4900000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only