missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Molybdate, For Silica, Certified, 10% (w/v), LabChem™
$115.27 - $504.60
Chemical Identifiers
| CAS | 12054-85-2 |
|---|---|
| Molecular Formula | H6MoNO2 |
| Molecular Weight (g/mol) | 148.00 |
| InChI Key | XCOMAVCHADVANQ-UHFFFAOYSA-O |
| Synonym | Molybdic acid ammonium salt tetrahydrate, Hexammonium heptamolybdate 4-hydrate |
| PubChem CID | 3084102 |
| IUPAC Name | ammonium hydrate molybdenum oxidandiide |
| SMILES | [NH4+].O.[O--].[Mo] |
Chemical Identifiers
| 12054-85-2 | |
| 148.00 | |
| Molybdic acid ammonium salt tetrahydrate, Hexammonium heptamolybdate 4-hydrate | |
| ammonium hydrate molybdenum oxidandiide |
| H6MoNO2 | |
| XCOMAVCHADVANQ-UHFFFAOYSA-O | |
| 3084102 | |
| [NH4+].O.[O--].[Mo] |
Specifications
| Colorless | |
| 9.69,87.91,2.4 | |
| (NH4)6Mo7O24·4H2O | |
| Molybdic acid ammonium salt tetrahydrate, Hexammonium heptamolybdate 4-hydrate | |
| XCOMAVCHADVANQ-UHFFFAOYSA-O | |
| ammonium hydrate molybdenum oxidandiide | |
| 3084102 | |
| 10% (w/v) | |
| Passes Test | |
| 1.02g/mL | |
| 1.1cSt | |
| 500 mL | |
| Ammonium Molybdate, For Silica |
| 12054-85-2,7732-18-5,1310-73-2 | |
| H6MoNO2 | |
| 1.02g/mL | |
| Soluble in water | |
| [NH4+].O.[O--].[Mo] | |
| 148.00 | |
| 1235.86 | |
| Certified | |
| Poly Bottle | |
| Passes Test | |
| Gaseous Ammonia | |
| Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Warning
Recommended Storage : Room Temperature