missing translation for 'onlineSavingsMsg'
Learn More

Ammonium molybdate, 99.99%, (trace metal basis)

CAS: 13106-76-8 | H8MoN2O4 | 196.01 g/mol

Supplier:  Thermo Scientific Chemicals 448540250

 View more versions of this product

Catalog No. AC448540250


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ammonium molybdate
25 g
H8MoN2O4
Solubility in water: soluble. Other solubilities: practically insoluble in alcohol
196.01
Trace Metal Basis
13106-76-8
99.99%
(NH4)2MoO4
196.01
99.99%
Glass bottle

Chemical Identifiers

13106-76-8
H8MoN2O4
196.01
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 236-031-3

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only