Learn More
Ammonium molybdate, 99.99% (metals basis)
CAS: 13106-76-8 | H8MoN2O4
$443.10 - $1380.47
Chemical Identifiers
| CAS | 13106-76-8 |
|---|---|
| Molecular Formula | H8MoN2O4 |
| MDL Number | MFCD00011596 |
Description
It is used to prepare phosphomolybdates, to measure the amount of phophates, silicates, arsenates, lead in pigments, river water and sea water. It is also used as a negative stain in electron microscopy, in photogrphy, ceramics, paints. It is used in the production of molybdenum. It is used in the preparation of dehydrogenation and desulphurization catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13106-76-8 | |
| H8MoN2O4 | |
| MFCD00011596 |
Specifications
| White | |
| Lump/Powder | |
| 99.99% (Metals basis) | |
| MFCD00011596 | |
| 196.02 | |
| Ammonium molybdate |
| 13106-76-8 | |
| 25 g | |
| H8MoN2O4 | |
| Soluble in water,acid and alkali,but insoluble in alcohol. | |
| Odorless |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves.
P264b-P270-P301+P312-P330-P501c
H302-H500
EINECSNumber : 236-031-3
RTECSNumber : QA4900000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only