missing translation for 'onlineSavingsMsg'
Learn More

Ammonium molybdate, 99.99% (metals basis)

Catalog No. AA4320614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4320614 25 g
AA4320622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4320614 Supplier Thermo Scientific Chemicals Supplier No. 04320614

CAS: 13106-76-8 | H8MoN2O4

It is used to prepare phosphomolybdates, to measure the amount of phophates, silicates, arsenates, lead in pigments, river water and sea water. It is also used as a negative stain in electron microscopy, in photogrphy, ceramics, paints. It is used in the production of molybdenum. It is used in the preparation of dehydrogenation and desulphurization catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13106-76-8
Molecular Formula H8MoN2O4
MDL Number MFCD00011596

Specifications

Color White
Physical Form Lump/Powder
Quantity 25 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information Soluble in water,acid and alkali,but insoluble in alcohol.
Formula Weight 196.02
Odor Odorless
Chemical Name or Material Ammonium molybdate
Hazard Category H302-H500
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 236-031-3
RTECSNumber QA4900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only