missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Molybdate, 1% (w/v) in 0.1 N Sulfuric Acid, Ricca Chemical

Catalog No. 660132
Change view
Click to view available options
Quantity:
1 L
Packaging:
Amber Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
660132 1 L Amber Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 660132 Supplier Ricca Chemical Company Supplier No. 660.132
Only null left

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9 , 12054-85-2
Molecular Formula H6MoNO2
Molecular Weight (g/mol) 148.00
InChI Key XCOMAVCHADVANQ-UHFFFAOYSA-O
Synonym Molybdic acid ammonium salt tetrahydrate, Hexammonium heptamolybdate 4-hydrate
PubChem CID 3084102
IUPAC Name ammonium hydrate molybdenum oxidandiide
SMILES [NH4+].O.[O--].[Mo]

Specifications

CAS Min % 0.96, 96.0, 0.98
CAS Max % 1.0, 100.0, 1.02
Physical Form Liquid
Quantity 1 L
Percent Purity 1% W/V
Grade Laboratory
Concentration or Composition (by Analyte or Components) 1% (w/v) in 0.1N Sulfuric Acid
Packaging Amber Poly Bottle
Chemical Name or Material Ammonium Molybdate
ShelfLife 12 months