missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Molybdate, 1% (w/v) in 0.1N Sulfuric Acid, Certified, LabChem™

Catalog No. LC112052
Change view
Click to view available options
Quantity:
1 L
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC112052 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC112052 Supplier LabChem Supplier No. LC112052

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ready-to-use analytical reagent solution for laboratory testing applications.

Chemical Identifiers

CAS 12054-85-2 , 7732-18-5 , 7664-93-9
Molecular Formula H6MoNO2
Molecular Weight (g/mol) 148.00
InChI Key XCOMAVCHADVANQ-UHFFFAOYSA-O
Synonym Molybdic acid ammonium salt tetrahydrate, Hexammonium heptamolybdate 4-hydrate
PubChem CID 3084102  
IUPAC Name ammonium hydrate molybdenum oxidandiide
SMILES [NH4+].O.[O--].[Mo]

Specifications

Color Colorless
CAS Max % 1, 98.51, 0.49
Physical Form Liquid
Quantity 1 L
Linear Formula (NH4)6Mo7O24·4H2O
Solubility Information Soluble in water
Formula Weight 1235.86
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Decomposition Information Sulfur compounds
Density 1g/mL
Chemical Name or Material Ammonium Molybdate, 1% (w/v) in 0.1N Sulfuric Acid
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature