missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium metavanadate, Puriss. p.a., Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™
Puriss. p.a., Reag. Ph. Eur.,≥99.5%
$2186.68 - $2186.68
Chemical Identifiers
| CAS | 7803-55-6 |
|---|---|
| Molecular Formula | H4NO3V |
| Molecular Weight (g/mol) | 116.98 |
| MDL Number | MFCD00011430 |
| InChI Key | YBVKNHXQSRDWAA-UHFFFAOYSA-M |
| Synonym | ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate |
| PubChem CID | 516859 |
| IUPAC Name | vanadiumoylol amine |
| SMILES | N.O[V](=O)=O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7803-55-6 | |
| 116.98 | |
| YBVKNHXQSRDWAA-UHFFFAOYSA-M | |
| 516859 | |
| N.O[V](=O)=O |
| H4NO3V | |
| MFCD00011430 | |
| ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate | |
| vanadiumoylol amine |
Specifications
| 7803-55-6 | |
| H4NO3V | |
| MFCD00011430 | |
| ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate | |
| N.O[V](=O)=O | |
| 116.98 | |
| 116.98g/mol | |
| puriss. p.a. | |
| 2.32g/cm3at 25°C |
| 1 kg | |
| NH4VO3 | |
| 2859 | |
| YBVKNHXQSRDWAA-UHFFFAOYSA-M | |
| vanadiumoylol amine | |
| 516859 | |
| ≥99.5% | |
| Plastic Bottle | |
| Ammonium Metavanadate, Reag. Ph. Eur. |
Safety and Handling
P260-P284-P301 + P310-P305 + P351 + P338-P310
H301-H315-H319-H330-H335
EINECSNumber : 232-261-3
RTECSNumber : YW0875000