missing translation for 'onlineSavingsMsg'
Learn More

Ammonium metavanadate, Puriss. p.a., Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™

Catalog No. 6000346
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000346 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6000346 Supplier Honeywell Chemicals Supplier No. 311531KG
Only null left

Puriss. p.a., Reag. Ph. Eur.,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7803-55-6
Molecular Formula H4NO3V
Molecular Weight (g/mol) 116.98
MDL Number MFCD00011430
InChI Key YBVKNHXQSRDWAA-UHFFFAOYSA-M
Synonym ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate
PubChem CID 516859
IUPAC Name vanadiumoylol amine
SMILES N.O[V](=O)=O

Specifications

Quantity 1 kg
Linear Formula NH4VO3
UN Number 2859
Formula Weight 116.98g/mol
Percent Purity ≥99.5%
Grade puriss. p.a.
Packaging Plastic Bottle
Density 2.32g/cm3at 25°C
Chemical Name or Material Ammonium Metavanadate, Reag. Ph. Eur.
Hazard Statement P260-P284-P301 + P310-P305 + P351 + P338-P310
Precautionary Statement H301-H315-H319-H330-H335
EINECSNumber 232-261-3
RTECSNumber YW0875000