missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(III) sulfate dodecahydrate, MilliporeSigma™

Catalog No. M1037761000
Change view
Click to view available options
Quantity:
1 kg
5 kg
50 kg
Packaging:
Fibre carton
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1037761000 1 kg Plastic Bottle
M1037765000 5 kg Plastic Bottle
M1037769050 50 kg Fibre carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1037761000 Supplier Technidata Supplier No. 1.03776.1000

For analysis EMSURE™ ACS, ISO, Reag. Ph Eur

Chemical Identifiers

CAS 7783-83-7
Molecular Formula FeH28NO20S2
Molecular Weight (g/mol) 482.176
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
Synonym ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
PubChem CID 197097
IUPAC Name azanium;iron(3+);disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]

Specifications

Quantity 1 kg
Solubility Information Soluble in water: 1240g/L at 25°C
Formula Weight 482.19
Odor Odorless
Packaging Plastic Bottle
Chemical Name or Material Ammonium iron(III) sulfate dodecahydrate
EINECSNumber 233-382-4
Recommended Storage Store below 15°C (59°F), Tightly closed. Dry. Protected from Light.