missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(III) sulfate dodecahydrate, ACS, 98.5-102.0%

Catalog No. p-4407224
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3939136 500 g
AA39391A4 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3939136 Supplier Thermo Scientific Chemicals Supplier No. 03939136
Only null left

CAS: 7783-83-7 | FeH28NO20S2 | 482.176 g/mol

Ammonium iron(III) sulfate dodecahydrate is used in dyeing and printing textiles, as analytical lab reagent. It is often employed as an analytical standard, and has been used in a variety of other applications from nanomaterials to general redox reactions. It plays an important role for the inactivation of mycobacteriophage D29 bacteria which is vivale for viable mycobacterium smegmatis and m.tuberculosis. The crystalline ammonium ferrous sulfate has shown useful function as the high absorbed dose and low-temperature dosimeter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7783-83-7
Molecular Formula FeH28NO20S2
Molecular Weight (g/mol) 482.176
MDL Number MFCD00150004
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
Synonym ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
PubChem CID 197097
IUPAC Name azanium;iron(3+);disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]

Specifications

Melting Point 39°C to 41°C
Color Purple
Physical Form Broken Crystals
Quantity 500 g
Assay Percent Range 98.5-102.0%
Solubility Information Freely soluble in water and hydrochloric acid. Insoluble in alcohol.
Formula Weight 482.18 (266.00 Anhydrous)
Grade ACS
Odor Odorless
Appearance Pale purple broken crystals
Density 1.71 g/cm3
Chemical Name or Material Ammonium iron(III) sulfate dodecahydrate, For analysis ACS
Show More Show Less
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 233-382-4
RTECSNumber WS5900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only