missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iron(III) oxalate trihydrate, 98%

Catalog No. AAB2141136
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2141136 500 g
AAB2141122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2141136 Supplier Thermo Scientific Chemicals Supplier No. B2141136

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13268-42-3 | C6H18FeN3O15 | 428.06 g/mol

Ammonium iron(III) oxalate trihydrate is used in the preparation of Langmuir-Blodgett magnetic thin films. It is also involved in the production of blueprint paper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium iron(III) oxalate trihydrate is used in the preparation of Langmuir-Blodgett magnetic thin films. It is also involved in the production of blueprint paper.

Solubility
Soluble in water. Insoluble in ethanol.

Notes
Light sensitive and hygroscopic. Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 13268-42-3
Molecular Formula C6H18FeN3O15
Molecular Weight (g/mol) 428.06
MDL Number MFCD00150137
InChI Key ARJSTCBPHIPXIJ-UHFFFAOYSA-K
Synonym Ferric ammonium oxalate; Iron(III) ammonium oxalate
IUPAC Name triammonium hexaoxo-1,4,6,9,10,13-hexaoxa-5-ferraspiro[4.4⁵.4⁵]tridecane-5,5,5-tris(ylium) trihydrate
SMILES [NH4+].[NH4+].[NH4+].O.O.O.O=C1O[Fe+3]234(OC1=O)OC(=O)C(=O)O2.O=C(O3)C(=O)O4

Specifications

Quantity 500 g
Sensitivity Hygroscopic; Light sensitive
Density 1.78
UN Number UN3288
Merck Index 14,517
Solubility Information Soluble in water. Insoluble in ethanol.
Formula Weight 428.07 (374.02 Anhydrous)
Percent Purity 98%
Chemical Name or Material Ammonium iron(III) oxalate trihydrate
Hazard Category H302-H312-H314-H335
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 238-090-0
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only