missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Iron(Iii) Citrate, Honeywell™
Purum p.a., 14.5-16% Fe basis (RT), powder, green
Supplier: Honeywell Chemicals 097131KG
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
| Ammonium Iron(Iii) Citrate | |
| 1185-57-5 | |
| 1 kg | |
| C6H8O7 ·xFe3+ · yNH3 | |
| FRHBOQMZUOWXQL-UHFFFAOYSA-N | |
| azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+) | |
| 118984355 | |
| 14.5 to 16% Fe basis (RT) |
| Green | |
| Powder | |
| C6H11FeNO7+3 | |
| ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz | |
| C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3] | |
| 264.999 | |
| purum p.a. |
Chemical Identifiers
| 1185-57-5 | |
| 264.999 | |
| ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz | |
| azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+) |
| C6H11FeNO7+3 | |
| FRHBOQMZUOWXQL-UHFFFAOYSA-N | |
| 118984355 | |
| C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3] |
Safety and Handling
P261-P305 + P351 + P338
H315-H319-H335
EINECSNumber : 214-686-6
RTECSNumber : GE7540000