missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Iron(Iii) Citrate, Honeywell™

Catalog No. p-7071741
Change view
Click to view available options
Quantity:
50 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001863 50 g
6001862 500 g
6001861 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6001863 Supplier Honeywell Chemicals Supplier No. 0971350G
Only null left

Purum p.a., 14.5-16% Fe basis (RT), powder, green

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Chemical Identifiers

CAS 1185-57-5
Molecular Formula C6H11FeNO7+3
Molecular Weight (g/mol) 264.999
InChI Key FRHBOQMZUOWXQL-UHFFFAOYSA-N
Synonym ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz
PubChem CID 118984355
IUPAC Name azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+)
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3]

Specifications

Color Green
Physical Form Powder
Quantity 50 g
Linear Formula C6H8O7 ·xFe3+ · yNH3
Grade purum p.a.
Concentration or Composition (by Analyte or Components) 14.5 to 16% Fe basis (RT)
Chemical Name or Material Ammonium Iron(Iii) Citrate
Hazard Statement P261-P305 + P351 + P338
Precautionary Statement H315-H319-H335
EINECSNumber 214-686-6
RTECSNumber GE7540000